missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RARB Recombinant Rabbit Monoclonal Antibody (24GB1045)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA557689
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RARB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
Spécification
| RARB | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RARB | |
| A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; NR1B2; nuclear receptor subfamily 1 group B member 2; RAR beta; Rarb; RAR-b; RAR-beta; RARbeta1; RAR-Beta2; RAR-Beta4; RAR-epsilon; retinoic acid; retinoic acid receptor; retinoic acid receptor beta; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5915 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1045 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10826 | |
| RARB | |
| A synthesized peptide derived from human Retinoic Acid Receptor beta (aa 400-455). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu