Learn More
ACROBiosystems Rat CD3 epsilon&CD3 delta Heterodimer Protein, Llama Fc Tag&Llama Fc Tag (MALS verified)
Marque: ACROBiosystems CDD-R5257-100ug
Informations supplémentaires : Poids : 0.00010kg
Description
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Rat CD3E&CD3D Heterodimer Protein, Llama Fc Tag&Llama Fc Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 40.8 kDa (CD3E) and 40.9 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag at the C-terminus and subunit CD3D is fSpécification
NP_001101610.1/NP_037301.1 | |
100 μg | |
Gln 21 - Asp 103 (CD3E) & Phe 22 - Gly 106 (CD3D) | |
CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
40.8 kDa (CD3E) & 40.9 kDa (CD3D) | |
Rat CD3E&CD3D; Heterodimer Protein, Llama Fc Tag&Llama; Fc Tag (CDD-R5257) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Asp 103 (CD3E) & Phe 22 - Gly 106 (CD3D) (Accession # D4A5M2-1 (CD3E) & P19377-1 (CD3D)). | |
-20°C | |
CD3E & CD3D | |
Rat | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |