missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00€ - 496.00€
Spécification
| Antigène | PDGF-AA |
|---|---|
| Clone | 184805 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18790127
![]() |
R&D Systems
MAB1055-SP |
25 μg |
188.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18161241
![]() |
R&D Systems
MAB1055 |
500 μg |
496.00€
500µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Rat samples. It is validated for Western Blot.Spécification
| PDGF-AA | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| AAB59693 | |
| 5154 | |
| E. coli-derived recombinant rat PDGF-AA Ser87-Arg196 Accession # AAB59693 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, 5-35% cross-reactivity with recombinant rat (rr) PDGF-AB, recombinant human (rh) PDGF-AA, and rhPDGF-AB is observed and no cross-reactivity with rrPDGF-BB, rhPDGF-BB, or rhPDGF-CC is observed. |
| 184805 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| Pdgfa | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit