missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBP2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5145249
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RBP2 is an abundant protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. RBP2 may also modulate the supply of retinoic acid to the nuclei of endometrial cells during the menstrual cycle.
Spécification
| RBP2 | |
| Polyclonal | |
| Unconjugated | |
| RBP2 | |
| AA409370; C76986; Cellular retinol-binding protein II; CRABP-II; CRBP2; Crbp-2; CrbpII; CRBP-II; Histone demethylase JARID1A; Jarid1a; jumonji, AT rich interactive domain 1A (Rbp2 like); Jumonji, AT rich interactive domain 1A (RBP2-like); jumonji/ARID domain-containing protein 1A; KDM5A; lysine (K)-specific demethylase 5A; lysine demethylase 5A; lysine-specific demethylase 5A; RBBP2; RBBP-2; RBP2; Rbp-2; RBPC2; retinoblastoma binding protein 2; retinoblastoma-binding protein 2; retinol binding protein 2; retinol binding protein 2, cellular; retinol-binding protein 2; Retinol-binding protein 2 cellular; retinol-binding protein 2, cellular | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 19660, 24710, 407611, 5948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P06768, P50120, P50121, Q08652 | |
| RBP2 | |
| Recombinant Human RBP2 protein, Met1-Lys134 (Accession #P50120). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu