Learn More
RCN1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB500-256JF646
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RCN1 Polyclonal antibody specifically detects RCN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| RCN1 | |
| Polyclonal | |
| Rabbit | |
| 5954 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FLJ37041, FLJ55835, PIG20, proliferation-inducing gene 20, RCAL, RCN, reticulocalbin 1, EF-hand calcium binding domain, reticulocalbin-1 | |
| A synthetic peptide that maps to a region between residues 1 and 75 of human Reticulocalbin-1, EF-hand calcium binding domain using the numbering given in Swiss-Prot entry Q15293 (GeneID 5954). |