Learn More
enQuireBio™ Recombinant Bovine aFGF Protein
A cDNA sequence encoding the aFGF was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10508-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
281160 | |
Greater than 90%. | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
Native Protein | |
MAEGETTTFT ALTEKFNLPL GNYKKPKLLY CSNGGYFLRI LPDGTVDGTK DRSDQHIQLQ LCAESIGEVY IKSTETGQFL AMDTDGLLYG SQTPNEECLF LERLEENHYN TYISKKHAEK HWFVGLKKNG RSKLGPRTHF GQKAILFLPL PVSSD |
aFGF Protein | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Stimulates growth of bovine capillary endothelial cells by 3-5 fold over 5% calf serum at 10-25ng/ml FGF and 0ug/ml heparin. | |
Bovine | |
Untagged | |
Each 5microg aFGF were lyophilized from 0.5ml solution containing 1mM sodium phosphate, pH 7 after filtration over a low binding membrane. |