Learn More
enQuireBio™ Recombinant Bovine BTC Protein
A cDNA sequence encoding the BTC was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10540-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
280737 | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
BTC | |
Bovine | |
Untagged | |
The Betacellulin Bovine Recombinant was lyophilized after extensive dialysis against 50mM acetic acid. |
BTC Protein | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARAAPGSGA SPLPLLPALA LGLVILHCVV ADGNSTRSPE DDGLLCGDHA ENCPATTTQP KRRGHFSRCP KQYKHYCIKG RCRFVVAEQT PSCVCDEGYA GARCERVDLF YLRGDRGQIL VICLIAVMVI FIILVVSICT CCHPLRKRRK RRKKEEEMET LGKDITPIND DIQETSIA |