missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine CASQ2 Protein

A cDNA sequence encoding the CASQ2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11280-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15906708

  • 715.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

483134
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Canine
MKRTHLFIAG LYLLASCRAE EGLNFPTYDG KDRVVSLTEK NFKQVLKKYD VLCLYYHESV SSDKVAQKQF QLKEIVLELV AQVLEHKDIG FVMVDAKKEA KLAKKLGFDE EGSLYVLKGD RTIEFDGEFA ADVLVEFLLD LIEDPVEIIN SKLEVQAFER IEDQIKLIGF FKSEESEYYK AFEEAAEHFQ PYIKFFATFD KGVAKKLSLK MNEVDFYEPF MDEPIAIPDK PYTEEELVEF VKEHQRPTLR RLRPEDMFET WEDDLNGIHI VAFAERSDPD GYEFLEILKQ VARDNTDNPD LSIVWIDPDD FPLLVAYWEK TFKIDLFKPQ IGVVNVTDAD SVWMEIPDDD DLPTAEELED WIEDVLSGKI NTEDDDNEEG DDGDDDEDDD DDDGNNSDEE SNDDSDDDDE
Greater than 90% as determined by SDS-PAGE.
CASQ2 Protein
Research Use Only
CASQ2
Canine
Untagged
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 10mM Tris-HCl and 1mM EGTA.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions