Learn More
enQuireBio™ Recombinant Canine CASQ2 Protein
A cDNA sequence encoding the CASQ2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11280-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
483134 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CASQ2 | |
Canine | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 10mM Tris-HCl and 1mM EGTA. |
CASQ2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Native Protein | |
MKRTHLFIAG LYLLASCRAE EGLNFPTYDG KDRVVSLTEK NFKQVLKKYD VLCLYYHESV SSDKVAQKQF QLKEIVLELV AQVLEHKDIG FVMVDAKKEA KLAKKLGFDE EGSLYVLKGD RTIEFDGEFA ADVLVEFLLD LIEDPVEIIN SKLEVQAFER IEDQIKLIGF FKSEESEYYK AFEEAAEHFQ PYIKFFATFD KGVAKKLSLK MNEVDFYEPF MDEPIAIPDK PYTEEELVEF VKEHQRPTLR RLRPEDMFET WEDDLNGIHI VAFAERSDPD GYEFLEILKQ VARDNTDNPD LSIVWIDPDD FPLLVAYWEK TFKIDLFKPQ IGVVNVTDAD SVWMEIPDDD DLPTAEELED WIEDVLSGKI NTEDDDNEEG DDGDDDEDDD DDDGNNSDEE SNDDSDDDDE |