Learn More
enQuireBio™ Recombinant Canine IFNG Protein
A cDNA sequence encoding the IFNG was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12378-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
403801 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
IFNG | |
Canine | |
His | |
IFNG protein solution (0.5 mg/ml) contains 20mM MES (pH6.0), 20% glycerol, 0.1M NaCl and 1mM EDTA. |
IFNG Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQAMFFKE IENLKEYFNA SNPDVSDGGS LFVDILKKWR EESDKTIIQS QIVSFYLKLF DNFKDNQIIQ RSMDTIKEDM LGKFLNSSTS KREDFLKLIQ IPVNDLQVQR KAINELIKVM NDLSPRSNLR KRKRSQNLFR GRRASK |