Learn More
enQuireBio™ Recombinant Canine IL 3 Protein
A cDNA sequence encoding the IL 3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10713-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
481497 | |
Greater than 97.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
IL3 | |
Canine | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing 1X PBS pH 7.4. |
IL 3 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
RPFSTDLPKQ YFTMINEIME MLNKSPSPSE EPLDSNEKET LLEDTLLRPN LDVFLNASSK FHKNGLLIWN NLKEFLPLLP TPTPRGEPIS IMENNWGDFQ RKLKKYLEAL DNFLNFKNKP |