Learn More
enQuireBio™ Recombinant Chlamydia HSP70 Protein
A cDNA sequence encoding the Chlamydia HSP70 (Amino Acids 462-503) was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11411-EC-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
HSP70 Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged |
Chlamydia HSP70 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 8M Urea, 60 mM NaCl and 50% glycerol. |