Learn More
enQuireBio™ Recombinant E. coli fabF Protein
A DNA sequence encoding the Escherichia coli (strain K12) fabF, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7326-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_415613.1 | |
fabF Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946665 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
fabF | |
Recombinant Protein | |
E. coli | |
SKRRVVVTGL GMLSPVGNTV ESTWKALLAG QSGISLIDHF DTSAYATKFA GLVKDFNCED IISRKEQRKM DAFIQYGIVA GVQAMQDSGL EITEENATRI GAAIGSGIGG LGLIEENHTS LMNGGPRKIS PFFVPSTIVN MVAGHLTIMY GLRGPSISIA TACTSGVHNI GHAARIIAYG DADVMVAGGA EKASTPLGVG GFGAARALST RNDNPQAASR PWDKERDGFV LGDGAGMLVL EEYEHAKKRG AKIYAELVGF GMSSDAYHMT SPPENGAGAA LAMANALRDA GIEASQIGYV NAHGTSTPAG DKAEAQAVKT IFGEAASRVL VSSTKSMTGH LLGAAGAVES IYSILALRDQ AVPPTINLDN PDEGCDLDFV PHEARQVSGM EYTLCNSFGF GGTNGSLIFK KI |