Learn More
enQuireBio™ Recombinant E. coli Trigger factor Protein
A DNA sequence encoding the Escherichia coli (strain K12) Trigger factor, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP9773-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
WP_001198385.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
tig | |
Recombinant Protein | |
Yeast | |
MQVSVETTQG LGRRVTITIA ADSIETAVKS ELVNVAKKVR IDGFRKGKVP MNIVAQRYGA SVRQDVLGDL MSRNFIDAII KEKINPAGAP TYVPGEYKLG EDFTYSVEFE VYPEVELQGL EAIEVEKPIV EVTDADVDGM LDTLRKQQAT WKEKDGAVEA EDRVTIDFTG SVDGEEFEGG KASDFVLAMG QGRMIPGFED GIKGHKAGEE FTIDVTFPEE YHAENLKGKA AKFAINLKKV EERELPELTA EFIKRFGVED GSVEGLRAEV RKNMERELKS AIRNRVKSQA IEGLVKANDI DVPAALIDSE IDVLRRQAAQ RFGGNEKQAL ELPRELFEEQ AKRRVVVGLL LGEVIRTNEL KADEERVKGL IEEMASAYED PKEVIEFYSK NKELMDNMRN VALEEQAVEA VLAKAKVTEK ETTFNELMN |
Trigger factor Protein | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |