Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Zaire GP Protein
A cDNA sequence encoding the Ebola Zaire GP was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11727-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Ebola Zaire GP Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
His |
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Zaire GP protein solution is supplied in Phosphate buffer with 25mM arginine and 0.02% sodium azide. |