Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Zaire Protein
A cDNA sequence encoding the Ebola Zaire Protein was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11728-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15918168
  • 905.00€ / 500µg
Expédition estimée: 04-07-2022
Spécification
Ebola Zaire Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Ebola Virus
His
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain).
Research Use Only
Recombinant Protein
E. coli
Ebola Zaire protein solution is supplied in Phosphate buffer and 0.02% sodium azide.