Learn More
enQuireBio™ Recombinant HIV-2 gp32 Protein
A cDNA sequence encoding the HIV-2 gp32 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12268-EC-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
HIV-2 gp32 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The HIV-2 gp32 solution (1 mg/ml) 0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol and 0.02% Sarcosyl. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |