missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACBD6 Protein

A cDNA sequence encoding the ACBD6 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10938-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15945628

  • 3595.00€ / 1mg
Expédition estimée: 08-03-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

ACBD6 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ACBD6 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMASSFLP AGAITGDSGG ELSSGDDSGE VEFPHSPEIE ETSCLAELFE KAAAHLQGLI QVASREQLLY LYARYKQVKV GNCNTPKPSF FDFEGKQKWE AWKALGDSSP SQAMQEYIAV VKKLDPGWNP QIPEKKGKEA NTGFGGPVIS SLYHEETIRE EDKNIFDYCR ENNIDHITKA IKSKNVDVNV KDEEGRALLH WACDRGHKEL VTVLLQHRAD INCQDNEGQT ALHYASACEF LDIVELLLQS GADPTLRDQD GCLPEEVTGC KTVSLVLQRH TTGKA
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions