missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-related protein 2/3 complex subunit 3, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8439-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001265485.1 | |
Actin-related protein 2 / 3 complex subunit 3 Protein | |
Research Use Only | |
ARPC3 | |
Recombinant Protein | |
E. coli | |
PAYHSSLMDP DTKLIGNMAL LPIRSQFKGP APRETKDTDI VDEAIYYFKA NVFFKNYEIK NEADRTLIYI TLYISECLKK LQKCNSKSQG EKEMYTLGIT NFPIPGEPGF PLNAIYAKPA NKQEDEVMRA YLQQLRQETG LRLCEKVFDP QNDKPSKWWT CFVKRQFMNK SLSG | |
Greater than 90% as determined by SDS-PAGE. |
10094 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |