missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adenine phosphoribosyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Adenine phosphoribosyltransferase, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5670-ec-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000476.1 | |
Adenine phosphoribosyltransferase Protein | |
Research Use Only | |
APRT | |
Recombinant Protein | |
E. coli | |
ADSELQLVEQ RIRSFPDFPT PGVVFRDISP VLKDPASFRA AIGLLARHLK ATHGGRIDYI AGLDSRGFLF GPSLAQELGL GCVLIRKRGK LPGPTLWASY SLEYGKAELE IQKDALEPGQ RVVVVDDLLA TGGTMNAACE LLGRLQAEVL ECVSLVELTS LKGREKLAPV PFFSLLQYE | |
Greater than 80% as determined by SDS-PAGE. |
353 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |