missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADFP Protein
A cDNA sequence encoding the ADFP was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10965-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
ADFP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS VAVDPQPSVV TRVVNLPLVS STYDLMSSAY LSTKDQYPYL KSVCEMAENG VKTITSVAMT SALPIIQKLE PQIAVANTYA CKGLDRIEER LPILNQPSTQ IVANAKGAVT GAKDAVTTTV TGAKDSVAST ITGVMDKTKG AVTGSVEKTK SVVSGSINTV LGSRMMQLVS SGVENALTKS ELLVEQYLPL TEEELEKEAK KVEGFDLVQK PSYYVRLGSL STKLHSRAYQ QALSRVKEAK QKSQQTISQL HSTVHLIEFA RKNVYSANQK IQDAQDKLYL SWVEWKRSIG YDDTDESHCA EHIESRTLAI ARNLTQQLQT TCHTLLSNIQ GVPQNIQDQA KHMGVMAGDI YSVFRNAASF KEVSDSLLTS SKGQLQKMKE SLDDVMDYLV NNTPLNWLVG PFYPQLTESQ NAQDQGAEMD KSSQETQRSEHKTH | |
Greater than 95% as determined by SDS PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Human ADFP was lyophilized from 0.5 mg/ml solution containing 20mM Tris pH 7.5, and 20mM NaCl. |