missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AK3L1 Protein

A cDNA sequence encoding the AK3L1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10990-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15925798

  • 205.00€ / 20µg
Expédition estimée: 04-07-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

205
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
AK4
Human
His
AK3L1 solution containing 20mM Tris-HCl pH 8.0, 2mM DTT and 20% glycerol.
AK3L1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASK LLRAVILGPP GSGKGTVSQR IAQNFGLQHL SSGHFLRENIKASTEVGEMA KQYIEKSLLV PDHVITRLMM SELENRRGQH WLLDGFPRTL GQAEALDKIC EVDLVISLNI PFETLKDRLS RRWIHPPSGR VYNLDFNPPH VHGIDDVTGE PLVQQEDDKP EAVAARLRQY KDVAKPVIEL YKSRGVLHQF SGTETNKIWP YVYTLFSNKI TPIQSKEAY
Vidéos
FDS
Documents
Certificats
Promotions

Promotions