missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKR1C4 Protein
A DNA sequence encoding the Homo sapiens (Human) AKR1C4, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5639-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001809.3 | |
AKR1C4 Protein | |
Research Use Only | |
AKR1C4 | |
Recombinant Protein | |
E. coli | |
MDPKYQRVEL NDGHFMPVLG FGTYAPPEVP RNRAVEVTKL AIEAGFRHID SAYLYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWCTFF QPQMVQPALE SSLKKLQLDY VDLYLLHFPM ALKPGETPLP KDENGKVIFD TVDLSATWEV MEKCKDAGLA KSIGVSNFNC RQLEMILNKP GLKYKPVCNQ VECHPYLNQS KLLDFCKSKD IVLVAHSALG TQRHKLWVDP NSPVLLEDPV LCALAKKHKQ TPALIALRYQ LQRGVVVLAK SYNEQRIREN IQVFEFQLTS EDMKVLDGLN RNYRYVVMDF LMDHPDYPFS DEY | |
Greater than 80% as determined by SDS-PAGE. |
1109 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |