missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AMIGO2 Protein
A cDNA sequence encoding the AMIGO2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11011-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
AMIGO2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VCPTACICAT DIVSCTNKNL SKVPGNLFRL IKRLDLSYNR IGLLDSEWIP VSFAKLNTLILRHNNITSIS TGSFSTTPNL KCLDLSSNKL KTVKNAVFQE LKVLEVLLLY NNHISYLDPS AFGGLSQLQK LYLSGNFLTQ FPMDLYVGRF KLAELMFLDV SYNRIPSMPM HHINLVPGKQ LRGIYLHGNP FVCDCSLYSL LVFWYRRHFS SVMDFKNDYT CRLWSDSRHS RQVLLLQDSF MNCSDSIING SFRALGFIHE AQVGERLMVH CDSKTGNANT DFIWVGPDNR LLEPDKEMEN FYVFHNGSLV IESPRFEDAG VYSCIAMNKQ RLLNETVDVT INVSNFTVSR SHAHEAFNTL EHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AMIGO2 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |