Learn More
enQuireBio™ Recombinant Human APOC1 Protein
A cDNA sequence encoding the APOC1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11049-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
341 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
APOC1 | |
Human | |
His | |
APOC1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |
APOC1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTPDVSSA LDKLKEFGNT LEDKARELIS RIKQSELSAK MREWFSETFQ KVKEKLKIDS |