missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARL14 Protein

A cDNA sequence encoding the ARL14 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11069-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15946048

  • 120.00€ / 5µg
Expédition estimée: 08-03-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

ARL14 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ARL14 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 40% glycerol, 200mM NaCl and 1mM EDTA.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLGSKNPQ TKQAQVLLLG LDSAGKSTLL YKLKLAKDIT TIPTIGFNVE MIELERNLSL TVWDVGGQEK MRTVWGCYCE NTDGLVYVVD STDKQRLEES QRQFEHILKN EHIKNVPVVL LANKQDMPGA LTAEDITRMF KVKKLCSDRN WYVQPCCALT GEGLAQGFRK LTGFVKSHMK SRGDTLAFFK QN
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions