missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL5A Protein
A cDNA sequence encoding the ARL5A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11073-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
ARL5A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGILFT RIWRLFNHQE HKVIIVGLDN AGKTTILYQF SMNEVVHTSP TIGSNVEEIV INNTRFLMWD IGGQESLRSS WNTYYTNTEF VIVVVDSTDR ERISVTREEL YKMLAHEDLR KAGLLIFANK QDVKECMTVA EISQFLKLTS IKDHQWHIQA CCALTGEGLC QGLEWMMSRL KIR | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ARL5A protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.2mM PMSF and 20% glycerol. |