missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Artemin / ARTN Protein
Recombinant Protein
Marque: enQuireBio™ QP5330-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001129687.1 | |
Artemin / ARTN Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG |
9048 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Artn | |
Human | |
Untagged | |
10 mM sodium citrate, 25 mM sodium chloride, pH 4.5 |