Learn More
enQuireBio™ Recombinant Human ASB8 Protein
A cDNA sequence encoding the ASB8 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11082-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
140461 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
ASB8 | |
Human | |
His | |
The ASB8 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
ASB8 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSSMWY IMQSIQSKYS LSERLIRTIA AIRSFPHDNV EDLIRGGADV NCTHGTLKPL HCACMVSDAD CVELLLEKGA EVNALDGYNR TALHYAAEKD EACVEVLLEY GANPNALDGN RDTPLHWAAF KNNAECVRAL LESGASVNAL DYNNDTPLSW AAMKGNLESV SILLDYGAEV RVINLIGQTP ISRLVALLVR GLGTEKEDSC FELLHRAVGH FELRKNGTMP REVARDPQLC EKLTVLCSAP GTLKTLARYA VRRSLGLQYL PDAVKGLPLP ASLKEYLLLL E |