missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AURKA Protein

A cDNA sequence encoding the AURKA was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11112-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15996178

  • 137.00€ / 2µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

6790
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
AURKA
Human
His
The AURKA solution containing 20mM Tris-HCl buffer (pH 8.0), 0.5mM DTT, 100mM NaCl, 0.1mM EDTA, 0.1mM EGTA, 0.1mM PMSF and 20% glycerol.
AURKA Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDRSKENCIS GPVKATAPVG GPKRVLVTQQ FPCQNPLPVN SGQAQRVLCP SNSSQRIPLQ AQKLVSSHKP VQNQKQKQLQ ATSVPHPVSR PLNNTQKSKQ PLPSAPENNP EEELASKQKN EESKKRQWAL EDFEIGRPLG KGKFGNVYLA REKQSKFILALKVLFKAQLE KAGVEHQLRR EVEIQSHLRH PNILRLYGYF HDATRVYLIL EYAPLGTVYR ELQKLSKFDE QRTATYITEL ANALSYCHSK RVIHRDIKPE NLLLGSAGEL KIADFGWSVH APSSRRTTLC GTLDYLPPEM IEGRMHDEKV DLWSLGVLCY EFLVGKPPFE ANTYQETYKRISRVEFTFPD FVTEGARDLI SRLLKHNPSQ RPMLREVLEH PWITANSSKP SNCQNKESAS KQS
Vidéos
FDS
Documents
Certificats
Promotions

Promotions