missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL2A1 Protein
A cDNA sequence encoding the BCL2A1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11143-100ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
597 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTDCEF GYIYRLAQDY LQYVLQIPQP GSGPSKTSRV LQKVAFSVQK EVEKNLKSCL DNVNVVSVDT ARTLFNQVME KEFEDDIINW GRIVTIFAFE GILIKKLLRQ QIAPDVDTYK EISYFVAEFI MNNTGEWIRQ NGGWENGFVK KFEPKS | |
Greater than 95% as determined by SDS-PAGE. |
BCL2A1 Protein | |
Research Use Only | |
BCL2A1 | |
Human | |
His | |
The BCL2A1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |