Learn More
enQuireBio™ Recombinant Human BLMH Protein
A cDNA sequence encoding the BLMH was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10535-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
642 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
BLMH | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSSGLNSEK VAALIQKLNS DPQFVLAQNV GTTHDLLDIC LKRATVQRAQ HVFQHAVPQE GKPITNQKSS GRCWIFSCLN VMRLPFMKKL NIEEFEFSQS YLFFWDKVER CYFFLSAFVD TAQRKEPEDG RLVQFLLMNP ANDGGQWDML VNIVEKYGVI PKKCFPESYT TEATRRMNDI LNHKMREFCI RLRNLVHSGA TKGEISATQD VMMEEIFRVV CICLGNPPET FTWEYRDKDK NYQKIGPITP LEFYREHVKP LFNMEDKICL VNDPRPQHKY NKLYTVEYLS NMVGGRKTLY NNQPIDFLKK MVAASIKDGE AVWFGCDVGK HFNSKLGLSD MNLYDHELVF GVSLKNMNKA ERLTFGESLM THAMTFTAVS EKDDQDGAFT KWRVENSWGE DHGHKGYLCM TDEWFSEYVY EVVVDRKHVP EEVLAVLEQE PIILPAWDPM GALAE |
BLMH Protein | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: > 1,000 pmole/min/ug. Measured by the hydrolysis of Met-AMC at pH 7.5, at 37C. | |
Human | |
His | |
The BLMH protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl Buffer (pH 8.0) and 10% Glycerol. |