missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1QTNF1 Protein

A cDNA sequence encoding the C1QTNF1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11230-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15926558

  • 137.00€ / 2µg
Expédition estimée: 30-06-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

C1QTNF1 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Human C1QTNF1 was lyophilized from 50mM Acetate Buffer pH 4.
Greater than 95% as determined by SDS PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS RVPHVQGEQQ EWEGTEELPS PPDHAERAEE QHEKYRPSQD QGLPASRCLR CCDPGTSMYP ATAVPQINIT ILKGEKGDRG DRGLQGKYGK TGSAGARGHT GPKGQKGSMG APGERCKSHY AAFSVGRKKP MHSNHYYQTV IFDTEFVNLY DHFNMFTGKF YCYVPGLYFF SLNVHTWNQK ETYLHIMKNE EEVVILFAQV GDRSIMQSQS LMLELREQDQ VWVRLYKGER ENAIFSEELD TYITFSGYLVKHATEP
Vidéos
FDS
Documents
Certificats
Promotions

Promotions