missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1QTNF6 Protein

A cDNA sequence encoding the C1QTNF6 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11233-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15996558

  • 205.00€ / 10µg
Expédition estimée: 09-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

C1QTNF6 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
0.23 mg/ml in 30mM Acetate Buffer pH 4.
Greater than 95% as determined by SDS PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS TFDRAVASGC QRCCDSEDPL DPAHVSSASS SGRPHALPEI RPYINITILK GDKGDPGPMG LPGYMGREGP QGEPGPQGSK GDKGEMGSPG APCQKRFFAF SVGRKTALHS GEDFQTLLFE RVFVNLDGCF DMATGQFAAP LRGIYFFSLN VHSWNYKETY VHIMHNQKEA VILYAQPSER SIMQSQSVML DLAYGDRVWV RLFKRQRENA IYSNDFDTYI TFSGHLIKAEDD
Vidéos
FDS
Documents
Certificats
Promotions

Promotions