missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C4c Protein
A cDNA sequence encoding the C4c was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11240-10mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
C4c Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
The Human Complement C4c solution is in Sodium Phosphate buffer pH 7.0 containing 0.15M NaCl and 0.09% NaN3. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 99.0%. |