Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5880-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001276841.1 | |
Casein kinase I isoform epsilon Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
102800317 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CSNK1E | |
Recombinant Protein | |
E. coli | |
MELRVGNKYR LGRKIGSGSF GDIYLGANIA SGEEVAIKLE CVKTKHPQLH IESKFYKMMQ GGVGIPSIKW CGAEGDYNVM VMELLGPSLE DLFNFCSRKF SLKTVLLLAD QMISRIEYIH SKNFIHRDVK PDNFLMGLGK KGNLVYIIDF GLAKKYRDAR THQHIPYREN KNLTGTARYA SINTHLGIEQ SRRDDLESLG YVLMYFNLGS LPWQGLKAAT KRQKYERISE KKMSTPIEVL CKGYPSEFST YLNFCRSLRF DDKPDYSYLR QLFRNLFHRQ GFSYDYVFDW NMLKFGAARN PEDVDRERRE HEREERMGQL RGSATRALPP GPPTGATANR LRSAAEPVAS TPASRIQPAG NTSPRAISRV DRERKVSMRL HRGAPANVSS SDLTGRQEVS RIPASQTSVP FDHLGK |