Learn More
enQuireBio™ Recombinant Human CCL17 / TARC / SCYA17 Protein
Recombinant Protein
Marque: enQuireBio™ QP97-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_002978.1 | |
CCL17 / TARC / SCYA17 Protein | |
Research Use Only | |
CCL17 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6361 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S | |
Greater than 95% as determined by SDS-PAGE. |