Learn More
enQuireBio™ Recombinant Human CFL2 / cofilin 2 / ADF Protein
A DNA sequence encoding the Homo sapiens (Human) CFL2 / cofilin 2 / ADF, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5825-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001230574.1 | |
CFL2 / cofilin 2 / ADF Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1073 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
CFL2 | |
Recombinant Protein | |
E. coli | |
MASGVTVNDE VIKVFNDMKV RKSSTQEEIK KRKKAVLFCL SDDKRQIIVE EAKQILVGDI GDTVEDPYTS FVKLLPLNDC RYALYDATYE TKESKKEDLV FIFWAPESAP LKSKMIYASS KDAIKKKFTG IKHEWQVNGL DDIKDRSTLG EKLGGNVVVS LEGKPL |