Learn More
enQuireBio™ Recombinant Human CRYZL1 Protein
A cDNA sequence encoding the CRYZL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11526-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
CRYZL1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CRYZL1 solution containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.1M NaCl and 20% glycerol. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKGLYFQQSS TDEEITFVFQ EKEDLPVTED NFVKLQVKAC ALSQINTKLL AEMKMKKDLF PVGREIAGIV LDVGSKVSFF QPDDEVVGIL PLDSEDPGLC EVVRVHEHYL VHKPEKVTWT EAAGSIRDGV RAYTALHYLS HLSPGKSVLI MDGASAFGTI AIQLAHHRGA KVISTACSLE DKQCLERFRP PIARVIDVSN GKVHVAESCL EETGGLGVDI VLDAGVRLYS KDDEPAVKLQ LLPHKHDIIT LLGVGGHWVT TEENLQLDPP DSHCLFLKGA TLAFLNDEVW NLSNVQQGKY LCILKDVMEK LSTGVFRPQL DEPIPLYEAK VSMEAVQKNQ GRKKQVVQF |