missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CST6 Protein

A cDNA sequence encoding the CST6 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11545-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15977588

  • 189.00€ / 10µg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

CST6 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CST6 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRPQERMVGE LRDLSPDDPQ VQKAAQAAVA SYNMGSNSIY YFRDTHIIKA QSQLVAGIKY FLTMEMGSTD CRKTRVTGDH VDLTTCPLAA GAQQEKLRCD FEVLVVPWQN SSQLLKHNCV QM
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions