Learn More
enQuireBio™ Recombinant Human CTGF Protein
Recombinant Protein
Marque:  enQuireBio™ QP5373-20ug
Informations supplémentaires : Poids : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 15926009
  • 252.00€ / 20µg
Expédition estimée: 07-06-2024
pour voir le stock disponible 
Spécification
NP_001892.1
CTGF Protein
Research Use Only
CTGF
Recombinant Protein
E. coli
MGKKCIRTP K. ISKPIKFE LS. GCTSMKT YRA. KFCGVC TDGR. CCTPH RTTTL. PVEF KCPDGE. VMK KNMMFIK. TC ACHYNCPG. D NDIFESLYY.  RKMYGDMA
Greater than 90 as determined by SDS-PAGE.
1490
20 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Bioactivity of Human CTGF confirmed in HUVEC cell proliferation assay.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA