missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DHPS Protein
A cDNA sequence encoding the DHPS was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11664-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
DHPS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEGSLEREAP AGALAAVLKH SSTLPPESTQ VRGYDFNRGV NYRALLEAFG TTGFQATNFG RAVQQVNAMI EKKLEPLSQD EDQHADLTQS RRPLTSCTIF LGYTSNLISS GIRETIRYLV QHNMVDVLVT TAGGVEEDLI KCLAPTYLGE FSLRGKELRE NGINRIGNLL VPNENYCKFE DWLMPILDQM VMEQNTEGVK WTPSKMIARL GKEINNPESV YYWAQKNHIP VFSPALTDGS LGDMIFFHSY KNPGLVLDIV EDLRLINTQA IFAKCTGMII LGGGVVKHHI ANANLMRNGA DYAVYINTAQ EFDGSDSGAR PDEAVSWGKI RVDAQPVKVY ADASLVFPLL VAETFAQKMD AFMHEKNED | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DHPS solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl and 20% glycerol. |