Learn More
enQuireBio™ Recombinant Human DHRS9 Protein
A cDNA sequence encoding the DHRS9 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11666-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
DHRS9 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DHRS9 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMRKGKL KIEDITDKYI FITGCDSGFG NLAARTFDKK GFHVIAACLT ESGSTALKAE TSERLRTVLL DVTDPENVKR TAQWVKNQVG EKGLWGLINN AGVPGVLAPT DWLTLEDYRE PIEVNLFGLI SVTLNMLPLV KKAQGRVINV SSVGGRLAIV GGGYTPSKYA VEGFNDSLRR DMKAFGVHVS CIEPGLFKTN LADPVKVIEK KLAIWEQLSP DIKQQYGEGY IEKSLDKLKG NKSYVNMDLS PVVECMDHAL TSLFPKTHYA AGKDAKIFWI PLSHMPAALQ DFLLLKQKAE LANPKAV |