missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DPPA4 Protein
A cDNA sequence encoding the DPPA4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11699-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
DPPA4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLRGSAS STSMEKAKGK EWTSTEKSRE EDQQASNQPN SIALPGTSAK RTKEKMSIKG SKVLCPKKKA EHTDNPRPQK KIPIPPLPSK LPPVNLIHRD ILRAWCQQLK LSSKGQKLDA YKRLCAFAYP NQKDFPSTAK EAKIRKSLQK KLKVEKGETS LQSSETHPPE VALPPVGEPP ALENSTALLE GVNTVVVTTS APEALLASWA RISARARTPE AVESPQEASG VRWCVVHGKS LPADTDGWVH LQFHAGQAWV PEKQEGRVSA LFLLPASNFP PPHLEDNMLC PKCVHRNKVL IKSLQWE | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DPPA4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |