Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase parkin Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase parkin, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6466-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_004553.2 | |
E3 ubiquitin-protein ligase parkin Protein | |
Research Use Only | |
PARK2 | |
Recombinant Protein | |
E. coli | |
MIVFVRFNSS HGFPVEVDSD TSIFQLKEVV AKRQGVPADQ LRVIFAGKEL RNDWTVQNCD LDQQSIVHIV QRPWRKGQEM NATGGDDPRN AAGGCEREPQ SLTRVDLSSS VLPGDSVGLA VILHTDSRKD SPPAGSPAGR SIYNSFYVYC KGPCQRVQPG KLRVQCSTCR QATLTLTQGP SCWDDVLIPN RMSGECQSPH CPGTSAEFFF KCGAHPTSDK ETSVALHLIA TNSRNITCIT CTDVRSPVLV FQCNSRHVIC LDCFHLYCVT RLNDRQFVHD PQLGYSLPCV AGCPNSLIKE LHHFRILGEE QYNRYQQYGA EECVLQMGGV LCPRPGCGAG LLPEPDQRKV TCEGGNGLGC GFAFCRECKE AYHEGECSAV FEASGTTTQA YRVDERAAEQ ARWEAASKET IKKTTKPCPR CHVPVEKNGG CMHMKCPQPQ CRLEWCWNCG CEWNRVCMGD HWFDV | |
Greater than 90% as determined by SDS-PAGE. |
5071 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |