missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF2B2 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF2B2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5970-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_055054.1 | |
EIF2B2 Protein | |
Research Use Only | |
EIF2B2 | |
Recombinant Protein | |
E. coli | |
MPGSAAKGSE LSERIESFVE TLKRGGGPRS SEEMARETLG LLRQIITDHR WSNAGELMEL IRREGRRMTA AQPSETTVGN MVRRVLKIIR EEYGRLHGRS DESDQQESLH KLLTSGGLNE DFSFHYAQLQ SNIIEAINEL LVELEGTMEN IAAQALEHIH SNEVIMTIGF SRTVEAFLKE AARKRKFHVI VAECAPFCQG HEMAVNLSKA GIETTVMTDA AIFAVMSRVN KVIIGTKTIL ANGALRAVTG THTLALAAKH HSTPLIVCAP MFKLSPQFPN EEDSFHKFVA PEEVLPFTEG DILEKVSVHC PVFDYVPPEL ITLFISNIGG NAPSYIYRLM SELYHPDDHV L | |
Greater than 80% as determined by SDS-PAGE. |
8892 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |