Learn More
enQuireBio™ Recombinant Human FKBP4 Protein
A cDNA sequence encoding the FKBP4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10609-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
2288 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
FKBP4 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTAEEMKATE SGAQSAPLPM EGVDISPKQD EGVLKVIKRE GTGTEMPMIG DRVFVHYTGW LLDGTKFDSS LDRKDKFSFDLGKGEVIKAW DIAIATMKVG EVCHITCKPE YAYGSAGSPP KIPPNATLVF EVELFEFKGE DLTEEEDGGI IRRIQTRGEG YAKPNEGAIV EVALEGYYKDKLFDQRELRF EIGEGENLDL PYGLERAIQR MEKGEHSIVY LKPSYAFGSV GKEKFQIPPN AELKYELHLK SFEKAKESWE MNSEEKLEQS TIVKERGTVYFKEGKYKQAL LQYKKIVSWL EYESSFSNEE AQKAQALRLA SHLNLAMCHL KLQAFSAAIE SCNKALELDS NNEKGLFRRG EAHLAVNDFE LARADFQKVLQLYPNNKAAK TQLAVCQQRI RRQLAREKKL YANMFERLAE EENKAKAEAS SGDHPTDTEM KEEQKSNTAG SQSQVETEA |
FKBP4 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH 8.0 using chymotrypsin. | |
Human | |
His | |
The FKBP4 protein solution contains 20mM Tris-HCl, pH 8 and 10% glycerol. |