Learn More
enQuireBio™ Recombinant Human FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10620-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
FSH Protein | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
HEK 293 | |
FSH subunit alpha: APDVQDCPECTLQENPFFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKSFSH subunit beta: NSCELTNITIAIEKEECRFCISINTTWCAGYCYTRDLVYKDPARPKIQKTCTFKELVYETVRVPGCAHHADSLYTYPVATQCHCGKCDSDSTDCTVRGLGPSYCSFGEMKE |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml. | |
Human | |
Untagged | |
The recombinant FSH was lyophilized from a concentrated (1 mg/ml) solution containing PBS. |