Learn More
enQuireBio™ Recombinant Human FUT3 Protein
A cDNA sequence encoding the FUT3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11911-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
FUT3 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FUT3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRVSRDDA TGSPRAPSGS SRQDTTPTRP TLLILLWTWP FHIPVALSRC SEMVPGTADC HITADRKVYP QADTVIVHHW DIMSNPKSRL PPSPRPQGQR WIWFNLEPPP NCQHLEALDR YFNLTMSYRS DSDIFTPYGW LEPWSGQPAH PPLNLSAKTE LVAWAVSNWK PDSARVRYYQ SLQAHLKVDV YGRSHKPLPK GTMMETLSRY KFYLAFENSL HPDYITEKLW RNALEAWAVP VVLGPSRSNY ERFLPPDAFI HVDDFQSPKD LARYLQELDK DHARYLSYFR WRETLRPRSF SWALDFCKAC WKLQQESRYQ TVRSIAAWFT |