Learn More
enQuireBio™ Recombinant Human GAD1 Protein
A cDNA sequence encoding the GAD1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11917-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
2571 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
GAD1 | |
Human | |
His | |
The GAD1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
GAD1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASSTPS SSATSSNAGA DPNTTNLRPT TYDTWCGVAH GCTRKLGLKI CGFLQRTNSL EEKSRLVSAF KERQSSKNLL SCENSDRDAR FRRTETDFSN LFARDLLPAK NGEEQTVQFL LEVVDILLNY VRKTFDRSTK VLDFHHPHQL LEGMEGFNLE LSDHPESLEQ ILVDCRDTLK YGVRTGHPRF FNQLSTGLDI IGLAGEWLTS TANTNMPSDM RECWLLR |